KR2I TFF SYSTEM FB COMM 2C, WITH SCALE 1 * 1 items
Catalog # SPECSYR2-U60
Supplier: Spectrum Laboratories
KR2I TFF SYSTEM FB COMM 2C, WITH SCALE 1 * 1 items
Catalog # SPECSYR2-U60
Supplier: Spectrum Laboratories
Frequently Bought Together
Accessories for WTW meters
Supplier: WTW
ThermoMixer® C Thermomixer
Supplier: EPPENDORF
Chamber slides, C-Slide
Supplier: NANOENTEK
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
Digestion tubes for Kjeldahl
Supplier: GERHARDT
Accessories for pH Meter, peqMETER 1.14
Supplier: CONSORT
VWR® pH CO 2500L, pH/Conductivity/°C Bench Meter with Colour Graphic Touch Screen
Supplier: VWR Collection
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Supplier: VWR Collection
Accessories for Turb® 430
Supplier: WTW
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
UV TUBE REPLACEMENT UV-8 MODEL S 365nM 1 * 1 items
Supplier: BENDA KONRAD
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
Supplier: WTW
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Colorimetrics tubes for gas measurements
Supplier: GASTEC
Melting point and boiling point capillaries and tubes
Supplier: BUCHI
Hydrogen generators, Precision SL 100
Supplier: PEAK SCIENTIFIC
BOTTOM SHELF WITH FLAP SS 1 * 1 items
Supplier: Thermo Scientific
Accessories for empty columns XK series
Supplier: Cytiva