ContactUsLinkComponent
,,6 components: Fluorides (F-) 20mg/l Chlorites (ClO2-) 10mg/l Chlorides (Cl-) 1000mg/l Sulphates (SO42-) 1000mg/l Chlorates (ClO3-) 10mg/l Bromides (Br-) 10mg/l in Water ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 12 months. (6 month 1 * 100 mL
Catalog # CMPLRM101321L1
,,6 components: Fluorides (F-) 20mg/l Chlorites (ClO2-) 10mg/l Chlorides (Cl-) 1000mg/l Sulphates (SO42-) 1000mg/l Chlorates (ClO3-) 10mg/l Bromides (Br-) 10mg/l in Water ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 12 months. (6 month 1 * 100 mL
Catalog # CMPLRM101321L1
Frequently Bought Together