Furnace MuFFle with scale anD soFtware lt9/11/sw/c550 tMax1100 with liFt Door anD therMocouPle hole in the Furnace Door 1 * 1 items
Catalog # NABELW095H1ANOZ1
Supplier: NABERTHERM
Furnace MuFFle with scale anD soFtware lt9/11/sw/c550 tMax1100 with liFt Door anD therMocouPle hole in the Furnace Door 1 * 1 items
Catalog # NABELW095H1ANOZ1
Supplier: NABERTHERM
Frequently Bought Together
Magnetic stirring bars
Supplier: VELP SCIENTIFIC
Vortex mixer, PV-1
Supplier: GRANT INSTRUMENTS
Magnetic stirrer, ESP
Supplier: VELP SCIENTIFIC
Magnetic hotplate stirrer, RH basic 2 IKAMAG®
Supplier: IKA
Accessories for hotplates, VHP-C Series
Supplier: IKA
VWR® INCU-Line® IL 10, IL 23, IL 56, Digital Incubators
Supplier: VWR Collection
Vortex mixers, ZX4 and TX4
Supplier: VELP SCIENTIFIC
VWR® VS-CV, Magnetic Stirrers
Supplier: VWR Collection
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Accessories for magnetic hotplate stirrer RCT basic
Supplier: IKA
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
Magnetic hotplate stirrers, C-MAG HS 7 control
Supplier: IKA
Bunsen burner, cartridge type
Supplier: USBECK Laborgeräte
VWR®, Accessories for Analogue, Digital and Pulsing Vortex Shakers
Supplier: VWR Collection
DrySyn® heating blocks
Supplier: ASYNT
Accessories for mortar grinder, RM 200
Supplier: Retsch GmbH