ContactUsLinkComponent
Panthera Tec Pol Cloud Binocular Head Siedentopf Type, 25º Inclined With Built-in Digital Camera COMS 1/1.8 Still Resolution8.0MP(3840x2160)Live Resolution Full HD (1920x1080) HDMI, Ethernet And WI-FI Widefield Eyepieces uc-wf10x/20mm wi 1 * 1 items
Catalog # MOTI1100403300231
Supplier:  MOTIC
Panthera Tec Pol Cloud Binocular Head Siedentopf Type, 25º Inclined With Built-in Digital Camera COMS 1/1.8 Still Resolution8.0MP(3840x2160)Live Resolution Full HD (1920x1080) HDMI, Ethernet And WI-FI Widefield Eyepieces uc-wf10x/20mm wi 1 * 1 items
Catalog # MOTI1100403300231
Supplier:  MOTIC
Frequently Bought Together