Copper (II) sulphate 180 g/l in 5% H2SO4 CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 500 mL
Catalog # CMPLZ13074499.L5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together