Platinum-Cobalt (Hazen) 5 Colour Standard 1 * 100 mL
Catalog # REGEHAZ501
Supplier: REAGECON
Frequently Bought Together
AMMONIUM CAL SOLUTION 10MG/L 1 * 500 mL
Supplier: YSI ENVIRONMENTAL
3-Methylhexane 1 * 100 mg
Supplier: EHRENSTORFER
MULTI ELEMENT STD 75 BLANK OIL S-21:30 1 * 200 g
Supplier: LGC Standards PROMOCHEM
TETRABROMOBISPHENOL A-DIGLYCIDYL ETHER 1 * 25 mg
Supplier: EHRENSTORFER
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
REFRACTIVE INDEX (NOMINAL: 1.4941 @ 25°C), CERTIFIED @ 20, 25 & 30°C 1 * 1 items
Supplier: LGC Standards PROMOCHEM
Organic reference standards, Chloroparaffin C10-C13, Dr. Ehrenstorfer
Supplier: EHRENSTORFER
Chitosan, Reference Standard
Supplier: USP
Milrinone, Reference Standards
Supplier: USP
ACRYLAMIDE 1000µG/ML IN METHANOL 1 * 1 mL
Supplier: EHRENSTORFER
Antimony standard solution, for ICP, in 1% HNO₃ and Tartaric Acid, VHG
Supplier: LGC Standards PROMOCHEM
Cevimeline Related Compounds, Reference Standard
Supplier: USP