Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Platinum-Cobalt (Hazen) 50 Colour Standard 1 * 100 mL
Catalog # REGEHAZ5001
Supplier:
REAGECON
Certificates
Expand image
Platinum-Cobalt (Hazen) 50 Colour Standard 1 * 100 mL
Catalog # REGEHAZ5001
Supplier:
REAGECON
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Zinc Standard Solution
Supplier: WTW
Triafamone 1 * 50 mg
Supplier: EHRENSTORFER
4-Chloro-o-toluidine
Supplier: EHRENSTORFER
VWR®, Density standards referenced at 25 °C
Supplier: VWR Chemicals
Methyl Caproate, Reference Standard
Supplier: USP
Fenarimol certified 1 * 0,1 g
Supplier: EHRENSTORFER
Vardenafil Related Compounds, Reference Standard
Supplier: USP
Desmethoxy Omeprazole, Pharmaceutical Analytical Impurities
Supplier: USP
Polyethylene Terephthalate, Reference Standards
Supplier: USP
Rivastigmine, Reference Standard
Supplier: USP
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Fonofos certified 1 * 0,1 g
Supplier: EHRENSTORFER