Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]VIACHECK 100% VIABILITY CONTROL SING 1 * 75 ST
Catalog # POLYVC50BSS-75
Supplier:
POLYSCIENCES
Expand image
[EN]VIACHECK 100% VIABILITY CONTROL SING 1 * 75 ST
Catalog # POLYVC50BSS-75
Supplier:
POLYSCIENCES
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Serological pipettes
Supplier: Greiner Bio-One
Serological pipettes, Stripette™
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cell Strainers
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF