Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]VIACHECK 0% VIABILITY CONTROL SINGLE 1 * 75 ST
Catalog # POLYVC10BSS-75
Supplier:
POLYSCIENCES
Expand image
[EN]VIACHECK 0% VIABILITY CONTROL SINGLE 1 * 75 ST
Catalog # POLYVC10BSS-75
Supplier:
POLYSCIENCES
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals