ContactUsLinkComponent
6 components: Fluorides (F-) 25mg/l Nitrites (NO2-) 25mg/l Nitrates (NO3-) 250mg/l Phosphates (PO43-) 250mg/l Chlorides (Cl-) 250mg/l Sulphates (SO42-) 750mg/l in Water 1 * 250 mL
Catalog # CMPLRM097302L25
6 components: Fluorides (F-) 25mg/l Nitrites (NO2-) 25mg/l Nitrates (NO3-) 250mg/l Phosphates (PO43-) 250mg/l Chlorides (Cl-) 250mg/l Sulphates (SO42-) 750mg/l in Water 1 * 250 mL
Catalog # CMPLRM097302L25
Frequently Bought Together