[EN]OXIVIR PLUS NC 2X5L 1 * 2 Fl
DIVY101106358
: Diversey
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.0% USP, Ph. Eur.
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Toluene ≥99%, GPR RECTAPUR® for pathology
: VWR Chemicals
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals