ContactUsLinkComponent
L-MARK Syringe 100UL Fixed Needle, Black PTFE, 51mm Length, 22s Gauge, LC Tip PK6 1 * 6 items
Catalog # SGEALMK.2620719B6P
L-MARK Syringe 100UL Fixed Needle, Black PTFE, 51mm Length, 22s Gauge, LC Tip PK6 1 * 6 items
Catalog # SGEALMK.2620719B6P
Frequently Bought Together