ContactUsLinkComponent
L-MARK Gold Syringe 10UL Fixed Needle, PTFE, 51mm Length, 22s Gauge, LC Tip PK6 1 * 6 items
Catalog # SGEALMK.2606416C6P
L-MARK Gold Syringe 10UL Fixed Needle, PTFE, 51mm Length, 22s Gauge, LC Tip PK6 1 * 6 items
Catalog # SGEALMK.2606416C6P
Frequently Bought Together