15 components: Ca 5000mg/l Al 1000mg/l Na 1000mg/l K 500mg/l Mg 500mg/l Cu 100mg/l Fe 100mg/l Mn 100mg/l Zn 100mg/l Cd 5mg/l Cr 5mg/l Li 5mg/l Mo 5mg/l Ni 5mg/l Pb 5mg/l in HNO3 5% 1 * 100 mL
Catalog # CMPLRM097340L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together