ContactUsLinkComponent
Tip 100-1000µl FlexTop Wide Orifice Extended-Length Aerogard® Filtered Racked Sterile 89mm in length ZAP™ Premier 1 * 576 items
Catalog # ALPCZP1250CS
Supplier:  Alpha Laboratories
Tip 100-1000µl FlexTop Wide Orifice Extended-Length Aerogard® Filtered Racked Sterile 89mm in length ZAP™ Premier 1 * 576 items
Catalog # ALPCZP1250CS
Supplier:  Alpha Laboratories
Frequently Bought Together