Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
[EN]KIT BRUKER ESI SPRAYER COMPLETE 1 * 1 ST
BRUK8705536
:
BRUKER DALTRONIK
[EN]KIT BRUKER ESI SPRAYER COMPLETE 1 * 1 ST
BRUK8705536
:
BRUKER DALTRONIK
Frequently Bought Together
Acid Red 183 30% (dye content), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Buffer powder pillows
: Hach
Reagents for total nitrogen determination using Lovibond® MD photometers
: Lovibond
ETHANOL STANDARD (3,0MG/ML) 10X1ML 1 * 1 KIT
: DIAGNOSTIC SYSTEMS DIASYS
Anti-PD1 Rabbit Recombinant Multiclonal Antibody [clone: RM1033]
: Abcam
Test kits, drop count tests/
: Hach
Cuvette tests for water testing for WTW spectrophotometers and photometers
: WTW
Anti-GAPDH Rabbit Recombinant Multiclonal Antibody [clone: RM1050]
: Abcam
Test kits, digital titration method
: Hach