Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PrimeLab powder reagents: Can with 10g of PL Nitrite HR 1 1 * 5 g
Catalog # WTIDPLPOW10NITRIHR
Supplier:
WATER ID PRIMELAB
Expand image
PrimeLab powder reagents: Can with 10g of PL Nitrite HR 1 1 * 5 g
Catalog # WTIDPLPOW10NITRIHR
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 32% in aqueous solution, AnalaR NORMAPUR® analytical reagent, for nitrogen analysis
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals