Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PrimeLab powder reagents: Can with 10g of PL Chromate 1 1 * 10 g
Catalog # WTIDPLPOW10CHROMAT
Supplier:
WATER ID PRIMELAB
Expand image
PrimeLab powder reagents: Can with 10g of PL Chromate 1 1 * 10 g
Catalog # WTIDPLPOW10CHROMAT
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals