ContactUsLinkComponent
High Precision Tweezers - Antimagnetic/Anti-acid SS - tips: straight, pointed, ultra fine. Length 110mm 1 * 1 items
Catalog # OUTI0C-11-SA
Supplier:  OUTILS RUBIS
High Precision Tweezers - Antimagnetic/Anti-acid SS - tips: straight, pointed, ultra fine. Length 110mm 1 * 1 items
Catalog # OUTI0C-11-SA
Supplier:  OUTILS RUBIS
Frequently Bought Together