STANDARD AU 4MG/L IN HNO3 5% HCL 15% 1 * 500 mL
CMPLRM087309L5.2
Frequently Bought Together
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo
pH buffer solutions, NIST traceable, Orion™
: Thermo Orion
Picric acid 10 g/l in aqueous solution Reag. Ph. Eur. 1065801
: VWR Chemicals