ContactUsLinkComponent
Custom FAME Standard mit n-Heptane 24 months stability Storage: 10C or colder Shipping Conditions: Ambient Package: 2ml Felx tubes Methyl Palmitate (C16:0) CAS 112-39-0 Concentration mg/mL:1 Methyl stearate (C18:0) CAS 112-61-8 1. Concentration mg/m 1 * 5 ST
Catalog # REST562339_5
Supplier:  Restek
Custom FAME Standard mit n-Heptane 24 months stability Storage: 10C or colder Shipping Conditions: Ambient Package: 2ml Felx tubes Methyl Palmitate (C16:0) CAS 112-39-0 Concentration mg/mL:1 Methyl stearate (C18:0) CAS 112-61-8 1. Concentration mg/m 1 * 5 ST
Catalog # REST562339_5
Supplier:  Restek
Frequently Bought Together