50 TABLETS SULPHITE LR PHOTOMETER 1 * 50 items
Catalog # WTIDTBSPSULFTLR50
Supplier: WATER ID PRIMELAB
50 TABLETS SULPHITE LR PHOTOMETER 1 * 50 items
Catalog # WTIDTBSPSULFTLR50
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Supplier: VWR Chemicals
VWR®, Immersion Oil
Supplier: VWR Collection
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Colorimetric test kit, sulphite, TESTOVAL®
Supplier: Heyl GmbH & Co, Gebr.
COD photometric test kits, Test’n Tube
Supplier: Hach
Colorimetric rapid tests, ammonium, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Powder pillows, ammonia
Supplier: Hach
Test strips
Supplier: Hach