50 TABLETS MOLYBDATE HR N° 2 PHOTOMETER 1 * 50 items
WTIDTBSPMDH250
Frequently Bought Together
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
: VWR Chemicals
Test strips, peracetic acid, Dosatest
: VWR Chemicals
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Joint grease, K.W.S.
: POLLATH
VWR®, OCT Embedding matrix for frozen sections
: VWR Chemicals
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
: CUSTOM MADE CHEMICALS