Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
50 TABLETS DPD N° 1 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSPD150
Supplier:
WATER ID PRIMELAB
Expand image
50 TABLETS DPD N° 1 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSPD150
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR®, Immersion Oil
Supplier: VWR Collection
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS