Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
50 TABLETS SULPHITEST N° 1 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSHSULFHR150
Supplier:
WATER ID PRIMELAB
Expand image
50 TABLETS SULPHITEST N° 1 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSHSULFHR150
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide 30% (w/w) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Crystal violet solution
Supplier: VWR Chemicals
Nitrite VHR L, 500 ml
Supplier: Lovibond
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
Powder pillows, ammonia
Supplier: Hach
Hydrochloric acid 5 mol/l (5 N) for biopharmaceutical production
Supplier: VWR Chemicals