Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
100 POWDER-PILLOWS MOLYVER3 1 * 100 items
Catalog # WTIDPPPMOLYVER3100
Supplier:
WATER ID PRIMELAB
Expand image
100 POWDER-PILLOWS MOLYVER3 1 * 100 items
Catalog # WTIDPPPMOLYVER3100
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS