Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
50 POWDER-PILLOWS HYD.PEROX. HR PHOT. 1 * 50 items
Catalog # WTIDPPPHPHR50
Supplier:
WATER ID PRIMELAB
Expand image
50 POWDER-PILLOWS HYD.PEROX. HR PHOT. 1 * 50 items
Catalog # WTIDPPPHPHR50
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide 32% in aqueous solution, AnalaR NORMAPUR® analytical reagent, for nitrogen analysis
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Accessories
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach