Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REAGENT KIT FOR PRIMELAB ID96 (50 TESTS) 1 * 1 items
Catalog # WTIDPL96-KIT
Supplier:
WATER ID PRIMELAB
Expand image
REAGENT KIT FOR PRIMELAB ID96 (50 TESTS) 1 * 1 items
Catalog # WTIDPL96-KIT
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Joint grease, K.W.S.
Supplier: POLLATH
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach
Test kits, photometric, Chloride, LCK cuvette tests
Supplier: Hach
Nitrite VHR L, 500 ml
Supplier: Lovibond