Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
65ML PL PHOSPHATE HR 1 (65 TESTS) 1 * 65 mL
Catalog # WTIDPL65PPHR1
Supplier:
WATER ID PRIMELAB
Expand image
65ML PL PHOSPHATE HR 1 (65 TESTS) 1 * 65 mL
Catalog # WTIDPL65PPHR1
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Acetic acid 99% ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Reagents for DR800, DR900, DR2010, DR2500, DR2400, DR2800, DR3800, DR3900, DR5000, DR6000
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Starch indicator solution, MDB
Supplier: Hach