Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
65ML PL NITRITE HR 2 (108 TESTS) 1 * 65 mL
Catalog # WTIDPL65NITRITEHR2
Supplier:
WATER ID PRIMELAB
Expand image
65ML PL NITRITE HR 2 (108 TESTS) 1 * 65 mL
Catalog # WTIDPL65NITRITEHR2
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Test strips, water hardness, Dosatest
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test kits, photometric, Surfactants, nonionic, LCK cuvette tests
Supplier: Hach
Turbidity standards calibration kit for 2100Q, Stablcal®
Supplier: Hach