Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
65 ML PL ISOTHIAZOLINONE 2 (108 TESTS) 1 * 65 mL
Catalog # WTIDPL65ISOZ2
Supplier:
WATER ID PRIMELAB
Expand image
65 ML PL ISOTHIAZOLINONE 2 (108 TESTS) 1 * 65 mL
Catalog # WTIDPL65ISOZ2
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution, GPR RECTAPUR® low in nitrogen
Supplier: VWR Chemicals
Ammonia solution 20%, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, sulfite, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Gelex secondary standards kit for 2100 series turbidity meters
Supplier: Hach
Powder pillows, iron
Supplier: Hach
Colorimetric rapid tests, nitrite, VISOCOLOR®
Supplier: MACHEREY-NAGEL