Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
65ML PL HYD. PEROXIDE HR 2 (541 TESTS) 1 * 65 mL
WTIDPL65HYDHRP2
:
WATER ID PRIMELAB
65ML PL HYD. PEROXIDE HR 2 (541 TESTS) 1 * 65 mL
WTIDPL65HYDHRP2
:
WATER ID PRIMELAB
Frequently Bought Together
RHAMNOGALACTURONAN 1 * 5 g
: MEGAZYME
Reagents for copper determination using Lovibond® MD photometers
: Lovibond
Reagents for DEHA determination with Lovibond® MD photometers
: Lovibond
Acid Red 183 30% (dye content), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
REAGENT KIT FOR PRIMELAB ID46 (50 TESTS) 1 * 1 items
: WATER ID PRIMELAB
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
: Thermo Fisher Scientific