Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REAGENT KIT FOR PRIMELAB ID51 (50 TESTS) 1 * 1 items
Catalog # WTIDPL51-KIT
Supplier:
WATER ID PRIMELAB
Expand image
REAGENT KIT FOR PRIMELAB ID51 (50 TESTS) 1 * 1 items
Catalog # WTIDPL51-KIT
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Test strips
Supplier: Hach
Nitrite VHR L, 500 ml
Supplier: Lovibond