Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REAGENT KIT FOR PRIMELAB ID30(108 TESTS) 1 * 1 items
Catalog # WTIDPL30-KIT
Supplier:
WATER ID PRIMELAB
Expand image
REAGENT KIT FOR PRIMELAB ID30(108 TESTS) 1 * 1 items
Catalog # WTIDPL30-KIT
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Potassium hydroxide 47% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Anti-Rb (phospho S780) Rabbit Recombinant Multiclonal Antibody [clone: RP23040067]
Supplier: Abcam
Reagents for DR800, DR900, DR2010, DR2500, DR2400, DR2800, DR3800, DR3900, DR5000, DR6000
Supplier: Hach
KIT REAGENT AMTAX SC 1-100MG/L NH4-N 1 * 1 KIT
Supplier: Hach
ALKALI SOLUTIONS 1 * 500 mL
Supplier: Hach
Sodium carbonate, Test Solution (TS)
Supplier: CPACHEM
Test cassettes for urine
Supplier: SURESCREEN