Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
30ML PL HYD. PEROXIDE LR 2 (250 TESTS) 1 * 30 mL
WTIDPL30HYDLRP2
:
WATER ID PRIMELAB
30ML PL HYD. PEROXIDE LR 2 (250 TESTS) 1 * 30 mL
WTIDPL30HYDLRP2
:
WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
: CUSTOM MADE CHEMICALS
Test kits, photometric, ammonium, LCK cuvette tests
: Hach