30ML PL HYD. PEROXIDE LR 1 (250 TESTS) 1 * 30 mL
WTIDPL30HYDLRP1
Frequently Bought Together
Sodium hydroxide 50% in aqueous solution, GPR RECTAPUR®
: VWR Chemicals
NANOCOLOR® Photometric Tube Tests, COD
: MACHEREY-NAGEL
Sodium hydroxide 30% in aqueous solution, TECHNICAL
: VWR Chemicals
Chinese ink, blue, Q Path® stain for histology
: VWR Chemicals
Fast Red RC Salt, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Direct Red 23, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Acid Red 183 30% (dye content), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Urine test strips, Medi-Test
: MACHEREY-NAGEL