30 ML PL DISSOX 1 (75 TESTS) 1 * 30 mL
Catalog # WTIDPL30DO1
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Qualitative test paper phosphatesmo MI for phosphatase in milk
Supplier: MACHEREY-NAGEL
Ascorbic acid test strips, Dosatest
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Standards, calibration for LCK cuvettes, Addista
Supplier: Hach
Citric acid
Supplier: Hach
Powder pillows, chlorine, free and total
Supplier: Hach
Test kits, photometric, Chloride, LCK cuvette tests
Supplier: Hach