REAGENT KIT FOR PRIMELAB ID16 (50 TESTS) 1 * 1 items
Catalog # WTIDPL16-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID16 (50 TESTS) 1 * 1 items
Catalog # WTIDPL16-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test strips, peroxide, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ascorbic acid test strips, Dosatest
Supplier: VWR Chemicals
Test kits, photometric, Aluminium, LCK cuvette tests
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach