REAGENT KIT FOR PRIMELAB ID128(50 TESTS) 1 * 1 items
Catalog # WTIDPL128-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID128(50 TESTS) 1 * 1 items
Catalog # WTIDPL128-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
1-Hexanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
Joint grease, K.W.S.
Supplier: POLLATH
Kjeldahl tablets, Kjeltabs
Supplier: THOMPSON & CAPPER
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals