Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REAGENT KIT FOR PRIMELAB ID122(50 TESTS) 1 * 1 items
Catalog # WTIDPL122-KIT
Supplier:
WATER ID PRIMELAB
Expand image
REAGENT KIT FOR PRIMELAB ID122(50 TESTS) 1 * 1 items
Catalog # WTIDPL122-KIT
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Powder pillows, reagents for Hach instruments
Supplier: Hach