REAGENT KIT FOR PRIMELAB ID120(50 TESTS) 1 * 1 items
Catalog # WTIDPL120-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID120(50 TESTS) 1 * 1 items
Catalog # WTIDPL120-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Supplier: VWR Chemicals
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
VWR® Bouin's solution
Supplier: VWR Chemicals
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Schiff reagent for analysis of aldehydes
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond