REAGENT KIT FOR PRIMELAB ID105(50 TESTS) 1 * 1 items
Catalog # WTIDPL105-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID105(50 TESTS) 1 * 1 items
Catalog # WTIDPL105-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide 30% (w/w) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Kjeldahl Tablets
Supplier: VWR Chemicals
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Supplier: VWR Chemicals
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
Supplier: VWR Chemicals
Qualitative test paper phosphatesmo MI for phosphatase in milk
Supplier: MACHEREY-NAGEL
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Reagents for DEHA determination with Lovibond® MD photometers
Supplier: Lovibond
Reference standard kits for use with MD 100 and MD 200 photometers
Supplier: Lovibond
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach