50 X 25 TUBETESTS COD MR (0 - 1500 MG/L) 1 * 1.250 Vial
Catalog # WTIDCOD-80-MR-50
Supplier: WATER ID PRIMELAB
50 X 25 TUBETESTS COD MR (0 - 1500 MG/L) 1 * 1.250 Vial
Catalog # WTIDCOD-80-MR-50
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test kits, photometric, Nitrite, LCK cuvette tests
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach