Nitric Acid 0.01M (0.01N) CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months. ,,,, Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 500 mL
Catalog # CMPLZ14221043
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together