25 components: 100mg/l each of Be V Cr Mn Co As Mo Cd Sb Pb U B Al Ti Fe Cu Ni Zn Se Sr Ba Tl Sn Rb Cs in HNO3 5% 1 * 100 mL
Catalog # CMPLRM076299L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together