Self-laminating Polyester Labels for M710
Labels, rectangular and round, B-116, M7-116-461, Polyester, Permanent acrylic, White/Transparent, M710, 15,24W×41,28H×12,70Ø mm, 150 sets/box
BRDY311977
: Brady
Self-laminating polyester labels for M710 printer.
- Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
- Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.
- Printer model:M710
- Type:B-116
- Colour:White/Transparent
- Description:Labels, rectangular and round
- Dimensions:15,24W×41,28H×12,70Ø mm
- Material:Polyester
- Seq:08
- Model:M7-116-461
- Packed:150 sets/box
- Adhesive:Permanent acrylic