HighPurS-Calcium Carbonate (CaCO3)- min. 99.99% - Noncertified Conservation 36 months. (36 months after opening) Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 50 g
Catalog # CMPLSL021.50G
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together