ContactUsLinkComponent
EDTA CAS:60-00-4 EC:200-449-4 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1.000 mg
Catalog # CMPLSB24400.1G
EDTA CAS:60-00-4 EC:200-449-4 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1.000 mg
Catalog # CMPLSB24400.1G
Frequently Bought Together