0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
3 components: Au 1000mg/l Ni 500mg/l Co 500mg/l in HNO3 5% HCl TR 1 * 100 mL
CMPLRM092985L1
:
CUSTOM MADE CHEMICALS LAB
3 components: Au 1000mg/l Ni 500mg/l Co 500mg/l in HNO3 5% HCl TR 1 * 100 mL
CMPLRM092985L1
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Total Iron Test Strips, Dosatest, VWR®
: VWR Chemicals
UltraClear® for histology, J.T.Baker®
: Avantor
EOSIN RAL 555 1L 1 * 1 L
: RAL
NANOCOLOR® Photometric Tube Tests, Surfactants
: MACHEREY-NAGEL
Formalin 10%, Q Path®, buffered
: VWR Chemicals
Test kits, photometric, Nitrite, LCK cuvette tests
: Hach
EOSIN RAL 555 2.5L 1 * 2,5 L
: RAL