BOTTLE GLASS 1000ML AMBER 28MM NECK WITH FITTED PTFE LINED CAP 1 * 22 items
Catalog # 331-0118
Supplier: LAB3 OWN PRODUCTION
BOTTLE GLASS 1000ML AMBER 28MM NECK WITH FITTED PTFE LINED CAP 1 * 22 items
Catalog # 331-0118
Supplier: LAB3 OWN PRODUCTION
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR® Cooling Bricks
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Universal Pipette Tips
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Sodium acetate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Pipette Tips, Standard
Supplier: VWR Collection
Reagents for DEHA determination with Lovibond® MD photometers
Supplier: Lovibond
VWR® Reagent Bottles, Narrow Neck
Supplier: VWR Collection